missing translation for 'onlineSavingsMsg'
Learn More

KLF4 Antibody (4E6), DyLight 488, Novus Biologicals™

Product Code. 18428943 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428943 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428943 Supplier Novus Biologicals Supplier No. NBP121047G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KLF4 Monoclonal antibody specifically detects KLF4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF4
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 4E6
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Accession No. O43474
Gene Alias Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut)
Host Species Mouse
Immunogen Recombinant human KLF4 (1-170aa) purified from E. coli (NP_004226.3).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9314
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.