missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566743
This item is not returnable.
View return policy
Description
Immunogen sequence: LMFDPVPVKQE AMDPVSVSYP SNYMESMKPN KYGVIYSTPL PEKFFQTPEG LSHGIQMEPV DL Highest antigen sequence identity to the following orthologs - mouse 92%, rat 94%.
Krueppel-like factors (KLFs) comprise a family of evolutionarily conserved zinc finger-containing transcription factors with diverse regulatory functions in cell growth, proliferation, differentiation and embryogenesis. Individual members of the Sp1-like/KLF family can function either as activators or repressors, depending on which promoter they bind and the coregulators with which they interact. KLF6, also designated Zf9 or CPBP (core promoter-binding protein), and KLF3 are Kruppel-like zinc finger containing transcription factors. KLF6 is rapidly induced during hepatic stellate cell activation and transactivates a reporter gene driven by the collagen I promoter, suggesting KLF6 plays a role in the response to tissue injury. KLF3 may play a role in hematopoiesis.
Specifications
| KLF3 | |
| Polyclonal | |
| Unconjugated | |
| Klf3 | |
| 9930027G08Rik; AL024007; Basic krueppel-like factor; basic kruppel like factor; Basic Kruppel- like factor; basic Kruppel-like factor; BKLF; CACCC-box-binding protein BKLF; klf12; Klf3; Krueppel-like factor 3; kruppel like factor 12; Kruppel like factor 3; Kruppel-like factor 12; Kruppel-like factor 3 (basic); MGC48279; TEF-2; transcript ch138; zinc finger containing transcription factor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51274 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P57682 | |
| Klf3 | |
| Recombinant Human KLF3. Recombinant protein control fragment (Product #RP-107399). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction