missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF2 Antibody (CL14502) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26859APC
This item is not returnable.
View return policy
Description
KLF2 Monoclonal antibody specifically detects KLF2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| KLF2 | |
| Monoclonal | |
| APC | |
| Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor | |
| This antibody was developed using a synthetic peptide derived from Q9Y5W3, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL14502 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 10365 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction