missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550340
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total KLF17 protein.
KLF17 is a transcription repressor that binds to the promoter of target genes and prevents their expression. Acts as a negative regulator of epithelial-mesenchymal transition and metastasis in breast cancer. Specifically binds the 5'-CACCC-3' sequence in the promoter of ID1, a key metastasis regulator in breast cancer, and repress its expression. May be a germ cell-specific transcription factor that plays important roles in spermatid differentiation and oocyte development.
Specifications
| KLF17 | |
| Polyclonal | |
| Unconjugated | |
| KLF17 | |
| 7420700M05Rik; AA420409; AU043468; C85123; D4Ertd561e; FLJ40160; germ cell specific zinc finger protein; germ cell-specific zinc finger protein; Gzf; KLF17; Krueppel-like factor 17; Kruppel like factor 17; Kruppel-like factor 17; novel zinc-finger protein; Zfp393; Zinc finger protein 393; ZLF393; ZNF393 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 128209 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q5JT82 | |
| KLF17 | |
| Fusion protein corresponding to residues near the C terminal of human Kruppel-like factor 17. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction