missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF17 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149260
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
KLF17 is a transcription repressor that binds to the promoter of target genes and prevents their expression. Acts as a negative regulator of epithelial-mesenchymal transition and metastasis in breast cancer. Specifically binds the 5'-CACCC-3' sequence in the promoter of ID1, a key metastasis regulator in breast cancer, and repress its expression. May be a germ cell-specific transcription factor that plays important roles in spermatid differentiation and oocyte development.
Specifications
| KLF17 | |
| Polyclonal | |
| Unconjugated | |
| KLF17 | |
| 7420700M05Rik; AA420409; AU043468; C85123; D4Ertd561e; FLJ40160; germ cell specific zinc finger protein; germ cell-specific zinc finger protein; Gzf; KLF17; Krueppel-like factor 17; Kruppel like factor 17; Kruppel-like factor 17; novel zinc-finger protein; Zfp393; Zinc finger protein 393; ZLF393; ZNF393 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 128209 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q5JT82 | |
| KLF17 | |
| E. coli-derived recombinant human KLF17, Met1-Gln228. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction