missing translation for 'onlineSavingsMsg'
Learn More

KLF16 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18377525 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377525 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377525 Supplier Novus Biologicals Supplier No. NBP310885100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KLF16 Polyclonal specifically detects KLF16 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF16
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias basic transcription element binding protein 4, Basic transcription element-binding protein 4, BTEB4Transcription factor NSLP2, dopamine receptor regulating factor, DRRF, Krueppel-like factor 16, Kruppel-like factor 16, Novel Sp1-like zinc finger transcription factor 2, NSLP2BTE-binding protein 4, Transcription factor BTEB4
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human KLF16 (NP_114124). Peptide sequence GAAAAPHLLAASILADLRGGPGAAPGGASPASSSSAASSPSSGRAPGAAP
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83855
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.