missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF16 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | KLF16 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
KLF16 Polyclonal specifically detects KLF16 in Human samples. It is validated for Western Blot.Specifications
| KLF16 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| basic transcription element binding protein 4, Basic transcription element-binding protein 4, BTEB4Transcription factor NSLP2, dopamine receptor regulating factor, DRRF, Krueppel-like factor 16, Kruppel-like factor 16, Novel Sp1-like zinc finger transcription factor 2, NSLP2BTE-binding protein 4, Transcription factor BTEB4 | |
| The immunogen is a synthetic peptide directed towards the middle region of human KLF16 (NP_114124). Peptide sequence GAAAAPHLLAASILADLRGGPGAAPGGASPASSSSAASSPSSGRAPGAAP | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 83855 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title