missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF15 Antibody (1C10) [Allophycocyanin], Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42989APC
This item is not returnable.
View return policy
Description
KLF15 Monoclonal antibody specifically detects KLF15 in Human samples. It is validated for ELISA,Western Blot
Specifications
| KLF15 | |
| Monoclonal | |
| APC | |
| DKFZp779M1320, kidney-enriched Kruppel-like factor, KKLFKidney-enriched krueppel-like factor, Krueppel-like factor 15, Kruppel-like factor 15 | |
| Kruppel-like factor 15 - (Uniprot# Q9UIH9) | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 28999 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| 1C10 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction