missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107153
This item is not returnable.
View return policy
Description
Sequence homology: Dog: 100%; Human: 100%; Pig: 93%; Rabbit: 93%.
This gene encodes a member of the Kruppel-like family of transcription factors. The gene exhibits imprinted expression from the maternal allele in embryonic and extra-embryonic tissues. Expression of this gene is induced by TGF-beta, and the protein represses TGF-beta receptor II expression. The protein functions as part of a transcriptional co-repressor complex that also contains the transcriptional regulator SIN3A and histone deacetylase 2.
Specifications
| KLF14 | |
| Polyclonal | |
| Unconjugated | |
| KLF14 | |
| 5330411L03Rik; AL022736; Basic transcription element-binding protein 5; BTEB5; BTE-binding protein 5; KLF14; Krueppel-like factor 14; Kruppel like factor 14; Kruppel-like factor 14; transcription factor BTEB5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 136259 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q8TD94 | |
| KLF14 | |
| Synthetic peptide directed towards the N terminal region of human KLF14 (aa 2-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction