missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KLF13 Polyclonal antibody specifically detects KLF13 in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | KLF13 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | PE |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | basic transcription element binding protein 3, Basic transcription element-binding protein 3, BTEB3FKLF2, BTE-binding protein 3, Krueppel-like factor 13, Kruppel-like factor 13, Novel Sp1-like zinc finger transcription factor 1, NSLP1RFLAT1, RANTES factor of late activated T lymphocytes-1, RANTES factor of late activated T-lymphocytes 1, RFLAT-1FKLF-2, Sp1 like zinc finger transcription factor, Transcription factor BTEB3, Transcription factor NSLP1 |
| Host Species | Rabbit |
| Immunogen | Peptide corresponding to amino acids range from 27-56 of the Kruppel-like factor 13; transcription factor NSLP1; novel Sp1 like zinc finger transcription factor; RANTES factor of late activated T lymphocytes-1 [Homo sapiens]. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?