missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF13 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-23614APC
This item is not returnable.
View return policy
Description
KLF13 Polyclonal antibody specifically detects KLF13 in Mouse samples. It is validated for Western Blot
Specifications
| KLF13 | |
| Polyclonal | |
| Western Blot | |
| basic transcription element binding protein 3, Basic transcription element-binding protein 3, BTEB3FKLF2, BTE-binding protein 3, Krueppel-like factor 13, Kruppel-like factor 13, Novel Sp1-like zinc finger transcription factor 1, NSLP1RFLAT1, RANTES factor of late activated T lymphocytes-1, RANTES factor of late activated T-lymphocytes 1, RFLAT-1FKLF-2, Sp1 like zinc finger transcription factor, Transcription factor BTEB3, Transcription factor NSLP1 | |
| Peptide corresponding to amino acids range from 27-56 of the Kruppel-like factor 13; transcription factor NSLP1; novel Sp1 like zinc finger transcription factor; RANTES factor of late activated T lymphocytes-1 [Homo sapiens]. | |
| 0.1 mL | |
| Cancer, Chromatin Research, Transcription Factors and Regulators, Tumor Suppressors | |
| 51621 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction