missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF12 Mouse anti-Human, Clone: PCRP-KLF12-1E3, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £396.00
Specifications
| Antigen | KLF12 |
|---|---|
| Clone | PCRP-KLF12-1E3 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18394116
|
Novus Biologicals
NBP3-13848-20UG |
20 μg |
£197.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18326522
|
Bio-Techne
NBP3-13848-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLF12 Monoclonal antibody specifically detects KLF12 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
| KLF12 | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array | |
| Unconjugated | |
| Mouse | |
| DNA Replication Transcription Translation and Splicing | |
| 10 mM PBS with 0.05% BSA | |
| AP-2rep, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep | |
| Recombinant full-length human KLF12 protein (Uniprot: Q9Y4X4) | |
| Primary | |
| Store at 4°C. |
| PCRP-KLF12-1E3 | |
| Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q9Y4X4 | |
| 11278 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title