missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF12 Monoclonal Antibody (3D2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521324
This item is not returnable.
View return policy
Description
Peptide Sequence: MNIHMKRKTI KNINTFENRM LMLDGMPAVR VKTELLESEQ GSPNVHNYPD MEAVPLLLNN VKGEPPEDSL SVDHFQTQTE PVDLSINKAR.
Activator protein-2 alpha (AP-2 alpha) is a developmentally-regulated transcription factor and important regulator of gene expression during vertebrate development and carcinogenesis. The protein encoded by this gene is a member of the Kruppel-like zinc finger protein family and can repress expression of the AP-2 alpha gene by binding to a specific site in the AP-2 alpha gene promoter. Repression by the encoded protein requires binding with a corepressor, CtBP1. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| KLF12 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9Y4X4 | |
| KLF12 | |
| KLF12 (NP_009180, 1 a.a. ~ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3D2 | |
| Unconjugated | |
| KLF12 | |
| 2700063E05Rik; AI225908; AI315654; AP-2 repressor; AP2REP; AP-2rep; AP-2rep transcription factor; B130052C06Rik; BHLHB24; D530033K05Rik; HSPC122; ID; KLF12; KLF12 zinc finger transcriptional repressor; Krueppel-like factor 12; Kruppel like factor 12; Kruppel-like factor 12; transcriptional repressor AP-2rep | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11278 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction