missing translation for 'onlineSavingsMsg'
Learn More

KLF12 Antibody (PCRP-KLF12-1E3), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18383046 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383046 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383046 Supplier Novus Biologicals Supplier No. NBP314277AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KLF12 Monoclonal antibody specifically detects KLF12 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF12
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-KLF12-1E3
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias AP-2rep, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep
Host Species Mouse
Immunogen Recombinant full-length human KLF12 protein (Uniprot: Q9Y4X4)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 11278
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.