missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KLF12 Polyclonal antibody specifically detects KLF12 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | KLF12 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AP-2rep, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human Kruppel-Like Factor 12 using the numbering given in entry NP_009180.3 (GeneID 11278). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?