missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF12 Antibody (1230C), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB9097G
This item is not returnable.
View return policy
Description
KLF12 Monoclonal antibody specifically detects KLF12 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| KLF12 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| 1230C | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| AP-2rep, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep | |
| Mouse myeloma cell line NS0-derived human KLF12, Asn2-Val402, Accession # Q9Y4X4 | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 11278 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction