missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF11 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | KLF11 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18385054
|
Novus Biologicals
NBP3-15582-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18380327
|
Novus Biologicals
NBP3-15582-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLF11 Polyclonal antibody specifically detects KLF11 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| KLF11 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| FKLFFKLF1, Krueppel-like factor 11, Kruppel-like factor 11, MODY7TGFB-inducible early growth response protein 2, TIEG-2, TIEG2TGFB inducible early growth response 2, Tieg3, Transforming growth factor-beta-inducible early growth response protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 13-107 of human KLF11 (NP_003588.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Transcription Factors and Regulators | |
| PBS, 50% glycerol, pH7.3 | |
| 8462 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title