missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLF11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101058
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KLF11.
The protein encoded by this gene is a zinc finger transcription factor that binds to SP1-like sequences in epsilon- and gamma-globin gene promoters. This binding inhibits cell growth and causes apoptosis. Defects in this gene are a cause of maturity-onset diabetes of the young type 7 (MODY7). Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| KLF11 | |
| Polyclonal | |
| Unconjugated | |
| KLF11 | |
| 9830142A17; D12Ertd427e; FKLF; FKLF1; klf11; Krueppel-like factor 11; Kruppel like factor 11; Kruppel-like factor 11; MODY7; Tcfcp2l2; TGFB inducible early growth response 2; TGFB inducible early growth response 3; TGF-beta inducible early growth response protein 2; TGFB-inducible early growth response protein 2; TGFB-inducible early growth response protein 2b; TGFB-inducible early growth response protein 3; Tieg2; TIEG-2; Tieg2b; Tieg3; TIEG-3; transcription factor CP2-like 2; transforming growth factor-beta-inducible early growth response protein 2; Transforming growth factor-beta-inducible early growth response protein 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 194655, 8462 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14901, Q8K1S5 | |
| KLF11 | |
| A synthesized peptide derived from human KLF11(Accession O14901), corresponding to amino acid residues M1-T36. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction