missing translation for 'onlineSavingsMsg'
Learn More

KLF10 Antibody [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30735604 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735604 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735604 Supplier Novus Biologicals Supplier No. AF7016JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

KLF10 Polyclonal antibody specifically detects KLF10 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF10
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias EGRA, EGR-alpha, Krueppel-like factor 10, Kruppel-like factor 10, TGFB inducible early growth response, TGFB-inducible early growth response protein 1, TIEG-1, TIEG1TIEG, Transforming growth factor-beta-inducible early growth response protein 1
Host Species Sheep
Immunogen E. coli-derived recombinant human KLF10, Asn31-Ser154, Accession # Q13118
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 7071
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.