missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals KLF1 Antibody (4E10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00010661-M02
This item is not returnable.
View return policy
Description
KLF1 Monoclonal antibody specifically detects KLF1 in Human, Mouse samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| KLF1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EKLFerythroid-specific transcription factor EKLF, Erythroid krueppel-like transcription factor, erythroid Kruppel-like factor, HBFQTL6, INLU, Krueppel-like factor 1, Kruppel-like factor 1 (erythroid), monoclonal A3D8 | |
| KLF1 (NP_006554, 183 a.a. ~ 237 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PRTGLSVPAASGAPYGLLSGYPAMYPAPQYQGHFQLFRGLQGPAPGPATSPSFLS | |
| 0.1 mg | |
| Cancer | |
| 10661 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG3 κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 4E10 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_006554 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction