missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab Klerwipe Sterile Low Particulate Dry Wipe in VHP Packaging
Click to view available options
Dimensions (L x W):
100 x 100 mm
Description
- Low particulate binder free hydroentangled polyester/cellulose wipes
- Features Klertear which makes the packs easy to open even when wearing gloves
- Terminally sterilized by gamma irradiation
- Compatible with the Klercide range of Alcohols and Biocides
Specifications
Specifications
| Product Type | Dry Wipe |
| Length (Metric) | 100 mm |
| Width (Metric) | 100 mm |
| Material | Polyester Cellulose |
| Dimensions (L x W) | 100 x 100 mm |
| Format | Wipe |
| For Use With (Application) | Cleaning and Disinfection Procedures |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction