missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab™ Klerwipe Sporicidal Enhanced Peroxide Blended with DI Mop Wipe
Description
- BPR approved
- Ready-to-use disinfectant for proactive control of spores, particularly on larger surfaces where safe handling is especially important
- Sporicidal efficacy
- Short contact times against bacteria and fungi even in the presence of background soiling
- Low odor: Safe for operators and cleanroom surfaces
- Effective broad spectrum disinfectant efficacy even when the level of bioburden may be variable, such as during transfer disinfection
- No hazard classification: Free of aldehyde, chlorine and peracetic acid
- Broad virucidal efficacy
- Mop wipe format provides a practical solution for large surface disinfection in smaller cleanroom areas where bucket systems are difficult to use and create additional maintenance costs
- Easy to open peelable pack
- The product has low surface tension properties which ensure an optimum spread of the disinfectant and ideal surface wetting for effective results
- Certificates of Analysis and Sterility are available for every batch
- Full supporting technical documentation is available
Specifications
Specifications
| Product Type | Cleanroom Biocide |
| pH | 2.01 to 2.30 |
| Scent | Unscented |
| Sterility | Sterile |
| Appearance | Clear to Light Yellow |
| Composition | 1.5% Hydrogen Peroxide with Deionized Water |
| Dimensions (L x W) | 420 x 250 mm |
| Disposable | Single-use |
| Filtered | Filtered |
| Flammable | No |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?