missing translation for 'onlineSavingsMsg'
Learn More

Ecolab Klerwipe 70/30 IPA Tub Wipe Blended with Deionized Water

Product Code. p-200007131
Click to view available options
Basis Weight:
23 gsm
45 gsm
Includes:
15 x 100 wipes
15 x 200 wipes
Unit Size:
Pack of 1500
Pack of 3000
This item is not returnable. View return policy

Product Code. 17885497

Brand: Ecolab 6600990

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Klerwipe 70/30 IPA Tub wipes are sterile, non woven, polyester and cellulose blend wipes impregnated with 70% isopropyl alcohol and blended with deionized water.

  • BPR approved
  • For rapid controlled disinfection of small surfaces and equipment
  • Impregnated with 0.2 μm filtered IPA|DI before being terminally sterilised by gamma irradiated at not less than 25KG
  • Double bagged resealable tub with a low shedding aperture
  • Contains 200 wipes on a perforated roll
  • Wiping can improve the effectiveness of the cleaning and disinfection procedure. Klerwipe are highly absorbent and remain wet throughout use ensuring a consistent application of alcohol to the whole surface area
  • Using impregnated wipes eliminates over-spraying of alcohol, reducing the risk of inhalation, especially in tight working spaces. The alcohol evaporates completely and the wipes do not contain any chemical binders, so no undesirable residues remain
  • Certificates of Analysis are available with every batch

Specifications

Product Type Wipe
Length (Metric) 200 mm
Width (Metric) 230 mm
Material Polyester Cellulose
Basis Weight 23 gsm
Sterility Sterile
Includes 15 x 200 wipes
compliance-icons compliance-icons
Product Identifier
  • Klerwipe 70/30 IPA
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Isopropanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.