missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab™ Klerwipe™ 70/30 IPA, Sterile Wipes
Double packed in a convenient resealable tub for one-step sanitizing. Ecolab Klerwipe™ 70/30 IPA, Sterile Wipes are 200 x 230mm nonwoven wipes impregnated with 70% IPA (22g/m2).
Brand: Ecolab™ 3079370
This item is not returnable.
View return policy
Description
Sterile low particulate wipes are made from a binder free, hydroentangled polyester/cellulose blend. Each is Impregnated with 70% IPA. Easy open tubs (even when wearing gloves) feature Klertear technology.
- Synonyms: IPA wipes, isopropyl alcohol wipes
- IPA: C3H80, CAS 67-63-0, Molar Mass 60.1g/mol
- Low shedding aperture
- Suitable for Grade A, B and C/D
- Effective as a bactericidal and fungicidal agent (EN 1276, EN 1650, EN 13697)
Specifications
Bactericidal/Fungicidal Wetted Surface Wipes | |
Resealable Tub | |
15 Tubs ×, 200 Wipes | |
Sterile |
White | |
Sterile cleaning and sanitizing | |
22g/m2 of 70% isopropyl alcohol with deinonized water |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Klerwipe 70/30 IPA
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol