missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klerwipe™ 70/30 IPA Mop Wipe

Product Code. 15362488
Change view
Click to view available options
Dimensions (L x W):
420 x 250 mm
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15362488 420 x 250 mm Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15362488 Supplier Ecolab™ Supplier No. 3079350

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sterile low particulate wipes impregnated with 70% isopropyl alcohol blended with deionised water

  • The biocide blend is 0.2 micron filtered then filled and bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
  • A sterile ready-to-use daily disinfectant mop wipe, in a peelable pack, for large surfaces

Specifications

Description Mop Wipe
Dimensions (L x W) 420 x 250 mm
Packaging Quantity 10 x 20 Wipes
compliance-icons compliance-icons
Product Identifier
  • Klerwipe 70/30 IPA
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: Isopropyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.