missing translation for 'onlineSavingsMsg'
Learn More

Ecolab Klerwipe 70/30 IPA Mop Wipe Blended with Deionized Water

Product Code. 17875497
Change view
Click to view available options
Dimensions (L x W):
420 x 250 mm
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
17875497 420 x 250 mm Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17875497 Supplier Ecolab Supplier No. 6600979

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Klerwipe 70/30 IPA low particulate mop wipes are a sterile, ready-to-use daily disinfectant mop wipe, in a peelable pack, for large surfaces.

  • BPR approved
  • Rapid controlled disinfection of large surfaces
  • Ideal solution for high grade environments
  • 0.2 micron filtered then filled and bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
  • Fast and simple to use. The wipes are designed to fit easily onto the Klercide Sterile Mop Wipe Frames
  • 20 wipes per pack, reducing wastage at the end of a session
  • Klerwipe impregnated mop wipes eliminate the potential risk of mixing a concentrate or a dilution error and deliver the correct concentration of active ingredients to the surface being disinfected
  • Easy-open, bagged, peelable packs
  • No space consuming buckets or trolley system required
  • Certificates of Analysis are available for every batch

Specifications

Product Type Wipe
Length (Metric) 420 mm
Width (Metric) 250 mm
Material Polyester Cellulose
Dimensions (L x W) 420 x 250 mm
Sterility Sterile
compliance-icons compliance-icons
Product Identifier
  • Klerwipe 70/30 IPA
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: Isopropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.