missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klerwipe™ 70/30 Denatured Ethanol

Product Code. 15473720
Change view
Click to view available options
Dimensions (L x W):
420 x 250 mm
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15473720 420 x 250 mm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15473720 Supplier Ecolab™ Supplier No. 3079180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sterile low particulate mop wipes impregnated with 70% denatured ethanol blended with deionised water

  • The biocide blend is 0.2 micron filtered then filled and bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation.
  • A sterile ready-to-use daily disinfectant mop wipe, in a peelable pack, for large

Specifications

Description Low Particulate Mop Wipe
Dimensions (L x W) 420 x 250 mm
Packaging Quantity 10 x 20 Wipes
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.