missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klerwipe™ 70/30 Alcohol Impregnated Wipes

Product Code. 15396108
Change view
Click to view available options
Saturation Solution:
70/30 Alcohol
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Saturation Solution unitSize
15396108 70/30 Alcohol Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15396108 Supplier Ecolab™ Supplier No. 3079210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in rapid clean down of surfaces, equipment and production components. Ecolab™ Klerwipe™ 70/30 Alcohol Impregnated Wipes remain wet throughout use ensuring consistent application of alcohol to whole surface area.

  • Highly absorbent wipes
  • Ideal for sterile environments
  • Double bagged re-sealable pouch
  • Suitable for direct product contact use
  • 100% polyester or low particulate wipes
  • No wastage
  • 2 year shelf life

Certifications

Manufactured by a ISO 9001:2008, ISO 13485:2012 and European Medical Device Directive 93/42/EEC accredited company

Specifications

Product Type Impregnated Wipes
Absorbancy Highly absorbent
For Use With (Application) Rapid clean down of surfaces, equipment and production components
Saturation Solution 70/30 Alcohol
Sterility Sterile
compliance-icons
Product Identifier
  • Klerwipe 70/30 Denatured Ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.