missing translation for 'onlineSavingsMsg'
Learn More

Ecolab KlerReveal™ 26W Replacement Bulb

Product Code. 17526526
Change view
Click to view available options
Product Type:
Replacement Bulb
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type
17526526 Replacement Bulb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17526526 Supplier Ecolab Supplier No. 3056600
  • ONLINE EXCLUSIVE PRICE
    £153.30
    £139.28 / Pack of 1
    Save £14.02
    9% Off
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

KlerReveal™ 26W UV Replacement Bulb for use with KlerReveal™ Operator Training Kit.

  • 26W UV Bulb
  • Replacement bulb for KlerReveal™ Operator Training Kit
  • Used for correcting cleanroom techniques and processes by providing instant visual results

Specifications

Product Type Replacement Bulb
For Use With (Equipment) KlerReveal™ Operator Training Kit
Size 26 W
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.