missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klercide Sporicidal Low Residue Peroxide Blended with WFI Spray

Product Code. 17844544
Change view
Click to view available options
Container Type:
Trigger Spray
Unit Size:
6 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Container Type unitSize
17844544 Trigger Spray 6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17844544 Supplier Ecolab™ Supplier No. 6600535
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Klercide Sporicidal Low Residue Peroxide is a very low residue blend of high grade 6% hydrogen peroxide and water for injection. The biocide blend is 0.2 μm filtered and sterile filled under Grade A laminar flow, then triple bagged in a Grade B cleanroom.

The Sterishield Delivery System has been validated to enable an in-use shelf life of three months, so there is no need to discard unused product. Combining a quicker, more ergonomic application system with this low residue product, removes steps from the disinfection process therefore saving time and potential handling risk.

  • BPR approved
  • Endotoxin level guaranteed to be less than 0.25 EU/mL
  • Broad spectrum activity that includes proven effects against bacterial spores
  • Can be used in rotation with any other Ecolab™ biocide to proactively reduce bioburden levels including bacterial spores
  • Non-corrosive and triple bagged
  • Suitable for use on all types of hard surfaces in cleanrooms and controlled areas, isolators, LAFs, equipment and production vessels exteriors
  • Very low residue profile, reducing the need to perform rinsing steps after application
  • 1 L trigger spray incorporates the Sterishield Delivery System - a fully validated closed system designed to keep the fluid contents of the bottle sterile at all times to ensure full compliance with GMP Annex 1 requirements that disinfectant solutions are sterile throughout use
  • Certificates of Analysis and Sterility are available for every batch
  • Full supporting technical documentation is available

Specifications

Product Type Cleanroom Biocide
Form Clear Liquid
Scent Unscented
Sterility Sterile
Appearance Clear
Composition 6% Hydrogen Peroxide and Water for Injection
Container Type Trigger Spray
Disposable Single-use
Filtered Filtered
Flammable No
Quantity 6 x 1 L
Temperature Range (English) 0°C to 25°C
Show More Show Less
compliance-icons
Product Identifier
  • KLERCIDE SPORICIDAL LOW RESIDUE PEROXIDE
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Hydrogen peroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.