missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klercide Sporicidal Low Residue Peroxide Blended with WFI Capped

Product Code. 17874554
Click to view available options
Container Type:
Capped Bottle
Unit Size:
20 L
This item is not returnable. View return policy

Product Code. 17874554

Brand: Ecolab™ 6600543

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Klercide Sporicidal Low Residue Peroxide is a very low residue blend of high grade 6% hydrogen peroxide and water for injection. The biocide blend is 0.2 μm filtered and sterile filled under Grade A laminar flow, then triple bagged in a Grade B cleanroom.

  • BPR approved
  • Endotoxin level guaranteed to be less than 0.25 EU/mL
  • Broad spectrum activity that includes proven effects against bacterial spores
  • Can be used in rotation with any other Ecolab™ biocide or to pro-actively reduce bioburden levels including bacterial spores
  • Non-corrosive and triple bagged
  • 100% compatible with gassing isolators
  • Suitable for use on all types of hard surfaces in cleanrooms and controlled areas
  • Very low residue profile, reducing the need to perform rinsing steps after application
  • 5 L capped container for large surfaces
  • Certificates of Analysis and Sterility are available for every batch
  • Full supporting technical documentation is available

Specifications

Product Type Cleanroom Biocide
Form Clear Liquid
Scent Unscented
Sterility Sterile
Appearance Clear
Composition 6% Hydrogen Peroxide and Deionized Water
Container Type Capped Bottle
Disposable Single-use
Filtered Filtered
Flammable No
Quantity 4 x 5 L
Temperature Range (English) 0°C to 25°C
Show More Show Less
compliance-icons
Product Identifier
  • Klercide Sporicidal Low Residue Peroxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen peroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.