missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klercide Sporicidal Enhanced Peroxide Blended with DI Spray

Product Code. 17834544
Change view
Click to view available options
Container Type:
Trigger Bottle
Unit Size:
6 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Container Type unitSize
17834544 Trigger Bottle 6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17834544 Supplier Ecolab™ Supplier No. 6600520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Klercide Sporicidal Enhanced Peroxide is an enhanced blend of 1.5% hydrogen peroxide with deionized water that is sterile and ready-to-use. This biocide blend is 0.2 μm filtered then sterile filled and double bagged under Grade A laminar flow.

The Sterishield Delivery System is the only fully validated closed system designed to keep the fluid contents of the bottle sterile at all times. This ensures full compliance with GMP Annex 1 requirements that disinfectant solutions are sterile prior to use.

  • BPR approved
  • Ready-to-use disinfectant for proactive control of spores, particularly on larger surfaces where safe handling is especially important
  • Short contact times against bacteria and fungi even in the presence of background soiling
  • Low odor: Safe for operators and cleanroom surfaces
  • Effective broad spectrum disinfectant efficacy even when the level of bioburden may be variable, such as during transfer disinfection
  • No hazard classification: Free of aldehyde, chorine and peracetic acid
  • Broad virucidal efficacy
  • Supplied in a 1 L trigger spray incorporating the Sterishield Delivery System, which ensures in-use sterility for up to 3 months
  • Double bagged
  • Low surface tension properties which ensure an optimum spread of the disinfectant and ideal surface wetting for effective results
  • Certificates of Analysis and Sterility are available for every batch
  • Full supporting technical documentation is available

Specifications

Product Type Cleanroom Biocide
Form Liquid
pH 2.01 to 2.30
Scent Unscented
Sterility Sterile
Appearance Clear to Light Yellow
Composition 1.5% Hydrogen Peroxide with Deionized Water
Container Type Trigger Bottle
Disposable Single-use
Flammable No
Quantity 6 x 1 L
Temperature Range (English) 5°C to 25°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.