Learn More
Ecolab™ Klercide™ Sporicidal Active Chlorine
Description
- The biocide blend is 0.2 micron filtered and double bagged in a Grade C cleanroom.
- The product is a sterile and ready-to-use highly effective rotational sporicide for small and large surfaces.
Specifications
Specifications
| Sterility | Sterile |
| Description | Sterile Sporicidal Active Chlorine Capped |
| Quantity | 4 Pk. |
| Volume (Metric) | 5 L |
Safety and Handling
Product Identifier
- Klercide Sporicidal Active Chlorine
- Warning
- Substances and mixtures corrosive to metals Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H290-May be corrosive to metals.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- MIXTURE LIST-Contain: sodium hypochlorite