Learn More
Ecolab Klercide Hand Held Spray Adaptor
Product Code. 17556516
Change view
Click to view available options
Autoclavable:
Autoclavable
Product Code. Autoclavable
17556516 Autoclavable
1 options
Product Code. 17556516 Supplier Ecolab Supplier No. 10056356
  • ONLINE EXCLUSIVE PRICE
    £244.65
    £222.10 / Pack of 1
    Save £22.55
    9% Off
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Klercide Hand Held Spray Adapter has been designed for use with the Klercide 70/30 aerosol products.
  • Designed for cleanroom use with Klercide aerosol products
  • Allows the aerosol to be used in the same way was a trigger spray
  • Manufactured from stainless steel
  • Fully autoclavable
  • Designed for cleanroom use with Klercide aerosol products
  • Allows the aerosol to be used in the same way was a trigger spray
  • Manufactured from stainless steel
  • Minimizes cross contamination within the most critical environments
Specifications
Product Type Aerosol Adapter
Autoclavable Autoclavable
For Use With (Equipment) Klercide 70/30 Aerosol Products
Material Stainless Steel
Quantity 1 Unit