missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klercide™ Filtered Alcohol

Product Code. 15336068
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15336068 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15336068 Supplier Ecolab™ Supplier No. 3078200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in effective disinfection procedure. Ecolab™ Klercide™ Filtered Alcohol has fully adjustable trigger spray allowing liquid to be dispensed as either jet or spray enabling thorough wetting of surface.

  • Trigger sprays are available with lightweight options
  • Large droplet size reduces risk of inhalation
  • No wastage
  • Optimum surface coverage available as either 70% v/v Isopropyl Alcohol blended with deionised water or 70% v/v Denatured Ethanol blended with deionised water
  • Alcohol blend is 0.2 micron filtered, filled and double bagged in a Grade C (ISO Class 7) cleanroom
  • All spray bottles incorporate the patented SteriShield Delivery System and an adjustable trigger spray
  • Ready to use

Certifications

Manufactured by ISO 9001:2008, ISO 13485:2012 and European Medical Device Directive 93/42/EEC accredited company

Specifications

Product Type Filtered Alcohol
Composition 70% v/v Isopropyl Alcohol blended with deionised water or 70% v/v Denatured Ethanol blended with dei
Filtered Filtered
For Use With (Application) Disinfection procedure
Volume (Metric) 1 L
compliance-icons
Product Identifier
  • Klercide 70/30 Denatured Ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, 2-methylpropan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.