Learn More
Ecolab™ Klercide™ 70/30 Pharma Ethanol Blended With WFI
Description
- The alcohol blend is 0.2 micron filtered then filled and triple bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation.
Specifications
Specifications
| Description | Blended with WFI Spray |
| Quantity | 6/Cs. |
| Volume (Metric) | 1 L |
Safety and Handling
Product Identifier
- Klercide 70/30 Pharma Ethanol
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- MIXTURE LIST-Contains : ethanol, Isopropyl Alcohol