Learn More
Ecolab Klercide 70/30 Pharma Ethanol
Product Code. p-7225665
Click to view available options
Volume (Metric):
5 L
Product Code. 17556526
Brand: Ecolab 3078600
70% ethanol denatured with IPA blended with deionized water. The alcohol blend is 0.2 micron filtered then filled and double bagged in a Grade C cleanroom.
  • The alcohol blend is 0.2 micron filtered then filled
  • Double bagged in a Grade C cleanroom for ease of transfer
  • Simple and easy to apply with adjustable trigger spray
  • Equipped with the SteriShield Delivery System to ensure the integrity of the contents is preserved throughout use
Specifications
Sterility Non-sterile
Composition 70% Ethanol Denatured with Isopropyl Alcohol Blended with 30% Deionised Water
Volume (Metric) 5 L