Learn More
Ecolab Klercide 70/30 Pharma Ethanol
Product Code. 17596526
Change view
Click to view available options
Sterility:
Sterile
Product Code. Sterility
17596526 Sterile
1 options
Product Code. 17596526 Supplier Ecolab Supplier No. 3082550
Klercide 70/30 Pharma Ethanol is sterile ethanol with IPA as denaturant that is specified to offer a residue on evaporation comparable that that of pure ethanol EP. Highly effective for use in direct product contact areas. The product is formulated without non-volatile denaturants.
  • 0.2 micron filtered, then filled and triple bagged in a Grade C cleanroom before being terminally sterilized by gamma irradiation
  • Triple bagged and irradiated at no less than 25 kGy to ensure spore free alcohol
  • Ready-to-use directly on hard surfaces
  • Formulated without non-volatile denaturants
  • Leaves no surface residue contaminants such as Bitrex or MEK upon evaporation
Specifications
Sterility Sterile
Composition 70% Ethanol Denatured with IPA Blended with WFI