missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klercide™ 70/30 Isopropyl Alcohol Blended with WFI

Product Code. 15336088
Change view
Click to view available options
Volume (Metric):
500 mL
1 L
5 L
Unit Size:
Each
Pack of 12
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15336088 500 mL Pack of 12
15306088 5 L Pack of 4
16479139 5 L Each
15326088 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15336088 Supplier Ecolab™ Supplier No. 3078560

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

For use in effective disinfection procedure. Ecolab™ Klercide™ 70/30 Isopropyl Alcohol Blended with WFI is sterilized by gamma irradiation.

  • Available as either 70/30 Isopropyl Alcohol (IPA) or Denatured Ethanol blended with water for injection (WFI) or deionized water
  • Also available as 60% Isopropyl alcohol or Propan-1-ol and Denatured Ethanol, both blended with WFI
  • Use pharma grade alcohol in blend
  • Cleanroom processed, 0.2 micron filtered and terminally sterilized by gamma irradiation
  • Available as spray systems fitted with SteriShield Delivery System, which ensures sterility of contents is preserved throughout use, aerosols, stream aerosol or in 5L capped container
  • Available with complete supporting documentation

Specifications

Product Type Filtered Alcohol
Filtered Filtered
For Use With (Application) Disinfection procedure
Volume (Metric) 500 mL
compliance-icons compliance-icons
Product Identifier
  • Klercide 70/30 IPA
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Isopropyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.