Learn More
Ecolab™ Klercide™ 70/30 IPA Blended with WFI
Product Code. 15346088
Change view
Click to view available options
Description:
Blended with WFI Aerosol
Blended with WFI Stream Aerosol
Unit Size:
Pack of 24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
15346088 Blended with WFI Aerosol Pack of 24
15392438 Blended with WFI Stream Aerosol Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15346088 Supplier Ecolab™ Supplier No. 3078580
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
70% isopropyl alcohol blended with Water for Injection (WFI) with endotoxin level guaranteed to be less than 0.25 EU/mL
  • The alcohol blend is 0.2 micron filtered then filled and triple bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
Specifications
Description Blended with WFI Aerosol
Quantity 24/Pk.
Volume (Metric) 360 mL
compliance-icons compliance-icons
Product Identifier
  • Klercide 70/30 IPA Aerosol
Signal Word
  • Danger
Hazard Category
  • Flammable and non-flammable aerosols category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H222-Extremely flammable aerosol.
  • H229-Pressurised container: May burst if heated.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P211-Do not spray on an open flame or other ignition source.
  • P251-Do not pierce or burn, even after use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
Supplemental information
  • MIXTURE LIST-Contain: Isopropyl Alcohol