missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab Klercide 60/40 IPA
Description
- Filled and double bagged in a Grade C (ISO Class 7) cleanroom
- 0.2 micron filtered
- Spore free
- Endotoxin levels below 0.25 EU/mL
- Endotoxin tested to an approved pharmacopoeial method
- A sterile ready-to-use alcohol for filter testing
Specifications
Specifications
| Sterility | Sterile |
| For Use With (Application) | Testing Filter Membranes and Storing Filters |
| Volume (Metric) | 5 L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?