missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Klenow Fragment, LC (2 U/μL)

Product Code. 10450440 Shop All Thermo Scientific Products
Click to view available options
Quantity:
300 units
Unit Size:
300 units
This item is not returnable. View return policy

Product Code. 10450440

Brand: Thermo Scientific™ EP0054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Klenow Fragment is the Large Fragment of DNA Polymerase exhibiting 5'→3' polymerase, 3'→5' exonuclease (proofreading) activities, but lacking 5'→3' exonuclease activity.

Thermo Scientific Klenow Fragment is the large fragment of DNA polymerase I. It exhibits 5'→3' polymerase activity and 3'→5' exonuclease (proofreading) activity, but lacks 5'→3' exonuclease activity of DNA polymerase I.

Highlights

  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, aminoallyl-, biotin-, digoxigenin- and fluorescently-labeled nucleotides)
  • Active in restriction enzyme, PCR, RT, and T4 DNA Ligase buffers

Applications

  • DNA blunting by fill-in 3'-overhangs
  • Random-primed DNA labeling
  • Labeling by fill-in 5'-overhangs of dsDNA
  • DNA sequencing by the Sanger method
  • Site-specific mutagenesis of DNA with synthetic oligonucleotides
  • Second strand synthesis of cDNA
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
Polymerase DNA Polymerase I
Quantity 300 units
Product Type Klenow Fragment

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.