missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Klenow Fragment, exo– (5 U/μL)

Product Code. p-4521356 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1,500 units
300 units
Unit Size:
1500 units
300 units
This item is not returnable. View return policy

Product Code. 10669880

Brand: Thermo Scientific™ EP0421

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Klenow Fragment, exo- is the large fragment of DNA polymerase I. Exhibits 5'→3' polymerase activity, but lacks the 3'→5' and 5'→3' exonuclease activities.

Klenow Fragment, exo-, is the large fragment of DNA polymerase I . It exhibits 5'→3' polymerase activity, but lacks the 3'→5' and 5'→3' exonuclease activities of DNA Polymerase I. The 3'→5' exonuclease activity of the enzyme is eliminated by mutations in the 3'→5'-exonuclease active sit.

Highlights

  • Lacks 3'→5' exonuclease activity
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, fluorescein-, rhodamine-, aminoallyl-, biotin-labeled nucleotides)
  • Active in restriction enzyme, PCR, and RT buffers

Applications

  • Random-primed DNA labeling
  • Labeling by fill-in 5'-overhangs of dsDNA
  • Strand displacement amplification (SDA)
  • DNA sequencing by the Sanger method

Note

Klenow Fragment, exo- is not recommended for DNA blunting reactions prior to DNA ligation since it frequently adds one or more extra nucleotides to the 3'-terminus of blunt-end DNA substrates in a non-template directed fashion

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Polymerase DNA Polymerase I
Compatible Buffer Restriction Enzyme, PCR, RT Buffer
Quantity 300 units
Product Type Klenow Fragment

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.