missing translation for 'onlineSavingsMsg'
Learn More

KLC3 Rabbit anti-Human, Polyclonal , Abnova™

Rabbit polyclonal antibody raised against recombinant human KLC3.

Brand:  Abnova PAB29466.100uL

Product Code. 16157220

  • £535.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a member of the kinesin light chain gene family. Kinesins are molecular motors involved in the transport of cargo along microtubules, and are composed of two kinesin heavy chain (KHC) and two kinesin light chain (KLC) molecules. KLCs are thought to typically be involved in binding cargo and regulating kinesin activity. In the rat, a protein similar to this gene product is expressed in post-meiotic spermatids, where it associates with structural components of sperm tails and mitochondria. [provided by RefSeq]

Sequence: MSVQVAAPGSAGLGPERLSPEELVRQTRQVV
Specifications

Specifications

KLC3
Polyclonal
Rabbit polyclonal antibody raised against recombinant human KLC3.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
KLC3
Recombinant protein corresponding to human KLC3.
100 μL
Primary
Human
Liquid
Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:500-1:1000) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user.
KLC2/KLC2L/KLCt/KNS2B
Rabbit
Antigen affinity purification
RUO
147700
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.