missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563314
This item is not returnable.
View return policy
Description
Immunogen sequence: MSVQVAAPGS AGLGPERLSP EELVRQTRQV V Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
KLC3 encodes a member of the kinesin light chain gene family. Kinesins are molecular motors involved in the transport of cargo along microtubules, and are composed of two kinesin heavy chain (KHC) and two kinesin light chain (KLC) molecules. KLCs are thought to typically be involved in binding cargo and regulating kinesin activity. In the rat, a protein similar to this gene product is expressed in post-meiotic spermatids, where it associates with structural components of sperm tails and mitochondria.
Specifications
| KLC3 | |
| Polyclonal | |
| Unconjugated | |
| KLC3 | |
| BC017147; kinesin light chain 2; kinesin light chain 3; Kinesin light chain KLCt; KLC2; KLC2L; KLC2-like; Klc3; Klct; KNS2B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 147700 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P597 | |
| KLC3 | |
| Recombinant protein corresponding to Human KLC3. Recombinant protein control fragment (Product #RP-103260). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction