missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLC1 Recombinant Rabbit Monoclonal Antibody (23GB5925), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | KLC1 |
|---|---|
| Clone | 23GB5925 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088564
![]() |
Invitrogen™
MA552952 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087870
|
Invitrogen™
MA552953 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLC1 Recombinant Monoclonal antibody specifically detects KLC1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| KLC1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O88447, P37285, Q07866 | |
| 16593, 171041, 3831 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5925 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI874768; kinesin 2; kinesin 2 60/70kDa; kinesin light chain 1; Kinesin2; Klc; KLC 1; Klc1; KNS 2; KNS 2A; Kns2; KNS2A; medulloblastoma antigen MU-MB-2.50; MGC15245 | |
| A synthesized peptide derived from human KLC1 (501-573AA). | |
| Primary | |
| -20°C | |
| KLC1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title