missing translation for 'onlineSavingsMsg'
Learn More

KLC1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18405787 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405787 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405787 Supplier Novus Biologicals Supplier No. NBP146840AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KLC1 Polyclonal antibody specifically detects KLC1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLC1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias hKLC1B, hKLC1G, hKLC1J, hKLC1S, kinesin 2, kinesin 2 60/70kDa, kinesin light chain 1, KLC 1, KLChKLC1N, KNS2AhKLC1P, KNS2hKLC1R, medulloblastoma antigen MU-MB-2.50, MGC15245
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 523 and 573 of human kinesin light chain 1 using the numbering given in entry NP_891553.2 (GeneID 3831).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3831
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.