missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KL 001

Product Code. 30001647
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001647 50 mg 50mg
30001627 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001647 Supplier Tocris Bioscience™ Supplier No. 4685/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cryptochrome protein stabilizer; lengthens circadian period

Cryptochrome protein (CRY) stabilizer; blocks proteasome-mediated degradation of CRY1 and CRY2. Lengthens circadian period in mouse NIH-3T3 fibroblasts without altering casein kinase activity. Represses glucagon-induced hepatic gluconeogenesis through the induction of Pck1 and G6pc expression.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 309928-48-1
Molecular Formula C21H22N2O4S

Specifications

Quantity 50 mg
Formula Weight 398.48
Purity >98%
Molecular Weight (g/mol) 398.477
Chemical Name or Material KL 001
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.