missing translation for 'onlineSavingsMsg'
Learn More

Kjeltabs Cu-3.5, Foss™

Product Code. 10124264
Encompass
Change view
Click to view available options
Quantity:
1000 Tablets
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10124264 1000 Tablets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10124264 Supplier Foss Analytical Supplier No. 60046515

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Composition per Tablet: 3.5g K2SO4 + 0.4g CuSO4·5H2O

Specifications

Physical Form Flat Round Speckled Tablets
CAS 7758-99-8
Quantity 1000 Tablets
Molecular Formula CuH10O9S
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
IUPAC Name copper(2+) pentahydrate sulfate
Molecular Weight (g/mol) 249.68
PubChem CID 24463
ChEBI CHEBI:31440
Chemical Name or Material Copper Kjeltabs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.