missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kjeldahl Sodium Sulfate Catalyst Tablets, Technical, Contains 1g Na2SO4, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
:
1000tablets, Plastic powder jar
Description
1000tablets, Plastic powder jar
Specifications
Specifications
| Melting Point | 880°C |
| Color | White |
| Boiling Point | 1100°C |
| Packaging | Plastic powder jar |
| MDL Number | 3504 |
| Quantity | 1000 Tablets |
| Formula Weight | 142.04g/mol |
| Physical Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction